Polyacrylonitrile, white powder
Catalog # ICNA0521753805
Supplier: MP Biomedicals
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:5 g
- Seq:2210
Specifications
Specification Test Results
- Identity Test:Passes
Specification Test Results
About this item
:
Density:1.184 g/mL (25 °C) (lit.)
Storage Temperature:Ambient
CAS Number:25014-41-9
Formula:(C₃H₃N)n
MDL Number:MFCD00084395