Order Entry
Norway
ContactUsLinkComponent
 
Ethyl violet 80%
  ACRO410490250
 :  
undefined
Ethyl violet 80%
  ACRO410490250
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    25 g
  • Seq:
    3308

 

 

  • Appearance:
    Green Powder
  • Infrared spectrum:
    Conforms
  • Loss on drying:
    ≤4 %
  • Dye content:
    ≥75 %
  • UV Lambda max:
    593 to 599 nm (c=0.1, H2O)

 

 

 :
 :492.13 g/mol
 :Ambient
 :2390-59-2
 :C₃₁H₄₂ClN₃
 :MFCD00011765