Order Entry
Norway
ContactUsLinkComponent
VWR®, Support Rods
VWR®, Support Rods
Catalog # 444-0919
Leverandør:  VWR Collection
VWR®, Support Rods
Catalog # 444-0919
Leverandør:  VWR Collection
Spesifikasjoner
  • Ø:
    13 mm
  • Length:
    914 mm
  • Pk:
    1
Spesifikasjoner
Om dette produktet
Heavy-duty, top-of-the-line rods provide maximum strength and durability. Constructed of 303 stainless steel.