Organic reference standards, Vamidothion sulphone (NMIA P1730)
Catalog # PROONMIAP1730
Supplier: LGC Standards PROMOCHEM
CAS Number:
Specifications
- Pk:25 mg
- Standards:Reference standard
- CAS No.:70898-34-9
- Description:Vamidothion sulphone
- Formula:C₈H₁₈NO₆PS₂
- Description:Vamidothion sulphone
Specifications
About this item
Purity estimate obtained by subtraction from 100% of total impurities by HPLC with UV detection, thermogravimetric analysis, Karl Fischer analysis and 1H NMR analysis is 96,1 ± 4,0% mass fraction, (95% coverage interval). Supporting evidence is provided by elemental microanalysis and quantitative NMR using certified triphenylphosphine oxide as internal standard.
- White solid
- Moisture content ≤0,2% mass fraction
- Volatile residue ≤0,1% mass fraction
- Non volatile residue ≤0,4% mass fraction
- CAS no.: 70898-34-9
Synonym: O,O-dimethyl-S-[2-[(1-methyl-2-(methylamino)-2-oxoethyl) sulphonyl] ethyl] phosphorothioate.