Liquid acid cleaner, Citrajet®
Supplier: PerkinElmer
Certificates
About this item
Citrajet® cleaner is ideal for use in labware washers, manual and ultrasonic cleaning. Use on glassware, metals, plastics, ceramics, porcelain, rubber, berglass and suitable for acid washing and rinsing. Corrosion inhibiting formula removes metal oxides, scale, salts, inorganic residues, trace metals and amphoteric proteins.