Human plasma, type 2 diabetes mellitus
Catalog # PB006C-1-DM2
CAS Number:
Specifications
- Pk:2 mL
- Description:Plasma, type 2 diabetes mellitus, cryopreserved
Specifications
About this item
Human peripheral blood plasma used for researching the causes, diagnosis, supportive care and treatment of type 2 diabetes mellitus (DM 2). Collected from IRB-consented donors diagnosed with type 2 diabetes mellitus.
No pre-screening is done on fresh products, but donors are tested on day of collection for Hepatitis B, Hepatitis C, and HIV 1/2.
Storage: Store at −80 °C.
Caution: Intended for only research use and not for direct therapeutic or diagnostic use in human or animals, or for further manufacturing of therapeutic products or devices.