Megestrol Acetate, Reference Standards
USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.
: US Pharmacopea Reference Standard
USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.
: US Pharmacopea Reference Standard
- Catalog No:
- 1379106.
- 1379128.
- 1379139.
- 1379172.
- Description:
- Megestrol Acetate
- Megestrol Acetate Related Compound C
- Megestrol Acetate Related Compound D
- Megestrol Acetate Related Compound H
- Synonym(s):
- 6,17a-β-Dimethyl-3,17-dioxo-17a-homoandrosta-4,6-diene-17a-α-yl acetate
- 6-Methylene-3,20-dioxopregn-4-en-17-yl acetate
- CAS No.:
- 595-33-5
- 74910-22-8
- 32634-95-0
- 907193-65-1
- Formula:
- C₂₄H₃₂O₄
Product Family Options
- DescriptionStandardsPack typePk
- Megestrol AcetateUSP reference standardVial250 mg1 * 250 mgkr 4 600,00
Specifications
Cat. No.1379106.DescriptionMegestrol AcetateSynonym(s)CAS No.595-33-5FormulaC₂₄H₃₂O₄ - Megestrol Acetate Related Compound CUSP reference standardVial20 mg1 * 20 mgkr 5 850,00
Specifications
Cat. No.1379128.DescriptionMegestrol Acetate Related Compound CSynonym(s)6,17a-β-Dimethyl-3,17-dioxo-17a-homoandrosta-4,6-diene-17a-α-yl acetateCAS No.74910-22-8Formula - Megestrol Acetate Related Compound DUSP reference standardVial20 mg1 * 20 mgkr 8 900,00
Specifications
Cat. No.1379139.DescriptionMegestrol Acetate Related Compound DSynonym(s)6-Methylene-3,20-dioxopregn-4-en-17-yl acetateCAS No.32634-95-0Formula - Megestrol Acetate Related Compound HUSP reference standardVial20 mg1 * 20 mgkr 12 600,00
Specifications
Cat. No.1379172.DescriptionMegestrol Acetate Related Compound HSynonym(s)CAS No.907193-65-1Formula



