Medroxyprogesterone Acetate, Reference Standards
About this item
USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.
Certifications: US Pharmacopea Reference Standard
3 Options Available Below
Product Details & Documents
USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.
Certifications: US Pharmacopea Reference Standard
Specifications for Product Family
Specifications
Specifications
- Catalog No:
- 1378012.
- 1378023.
- 1378001.
- Description:
- Medroxyprogesterone Acetate Related Compound A
- Medroxyprogesterone Acetate Related Compound B
- Medroxyprogesterone Acetate
- Synonym(s):
- 6α-Methyl-3,20-dioxopregnan-17-yl acetate
- 17-Hydroxy-6α-methylpregn-4-ene-3,20-dione
- CAS No.:
- 69688-15-9
- 520-85-4
- 71-58-9
- Formula:
- C₂₄H₃₆O₄
- C₂₂H₃₂O₃
- C₂₄H₃₄O₄
Specifications
Product Family Options
Product Information
- StandardsPack typePkAvailabilityPrice
- 1378012.Medroxyprogesterone Acetate Related Compound AMedroxyprogesterone Acetate Related Compound AUSP reference standardVial25 mgLog in for availability1 * 25 mgkr 15 775,00Specifications:
More
Specifications
Cat. No.1378012.DescriptionMedroxyprogesterone Acetate Related Compound ASynonym(s)6α-Methyl-3,20-dioxopregnan-17-yl acetateCAS No.69688-15-9FormulaC₂₄H₃₆O₄ - 1378023.Medroxyprogesterone Acetate Related Compound BMedroxyprogesterone Acetate Related Compound BUSP reference standardVial25 mgLog in for availability1 * 25 mgkr 14 650,00Specifications:
More
Specifications
Cat. No.1378023.DescriptionMedroxyprogesterone Acetate Related Compound BSynonym(s)17-Hydroxy-6α-methylpregn-4-ene-3,20-dioneCAS No.520-85-4FormulaC₂₂H₃₂O₃ - 1378001.Medroxyprogesterone AcetateMedroxyprogesterone AcetateUSP reference standardVial200 mgLog in for availability1 * 200 mgkr 4 225,00Specifications:
More
Specifications
Cat. No.1378001.DescriptionMedroxyprogesterone AcetateSynonym(s)CAS No.71-58-9FormulaC₂₄H₃₄O₄
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



