Order Entry
Norway
ContactUsLinkComponent
Human recombinant Flagellin TLR5 Mutant (from E. coli)
Human recombinant Flagellin TLR5 Mutant (from E. coli)
Catalog # PRSI90-307
Leverandør:  ProSci Inc.
CAS-nummer:  
Human recombinant Flagellin TLR5 Mutant (from E. coli)
Catalog # PRSI90-307
Leverandør:  ProSci Inc.
CAS-nummer:  

Spesifikasjoner

  • Fp:
    10 µG
  • Protein/peptide type:
    Recombinant
  • Kilde:
    E. coli
  • Arter:
    Human
  • Tag sekvens:
    His Tag
  • Lagringsforhold:
    Stable for at least 6 months after receipt when stored at −20 °C.
  • Biologisk aktivitet:
    Does not activate TLR5 in human epithelial cell assays based on NF-kappaB luciferase fusions, in vivo on expression of CCL20 or IL-6. It can be used as negative control for TLR5 signaling assays.
  • Protein synonyms:
    FliC (TLR5 Mutant)
  • Protein/peptide name:
    Flagellin TLR5 Mutant
  • Renhet:
    >95% (SDS-PAGE)
  • Sekvens:
    1-495
  • Formulering:
    Lyophilized, Contains PBS.

Spesifikasjoner

Om dette produktet

Flagellin is the subunit protein which polymerises to form the filaments of bacterial flagella. It activates the innate immune system through the receptor Toll-like Receptor 5 (TLR5) or the intracellular NLRC4 protein. The Flagellin (TLR5 Mutant)(rec. ) cannot be detected by human and mouse TLR5. It has not been tested on NLRC4.

Fusion-Tag: His Tag

This recombinant proteins is for research use only.