Order Entry
Norway
ContactUsLinkComponent
Mouse recombinant FGF21 (from HEK293 cells)
Mouse recombinant FGF21 (from HEK293 cells)
Catalog # PRSI90-104
Supplier:  ProSci Inc.
CAS Number:  
Mouse recombinant FGF21 (from HEK293 cells)
Catalog # PRSI90-104
Supplier:  ProSci Inc.
CAS Number:  

Specifications

  • Pk:
    10 µG
  • Protein/peptide type:
    Recombinant
  • Source:
    HEK293 cells
  • Species:
    Mouse
  • Tag sequence:
    Fc Tag
  • Storage conditions:
    Working aliquots are stable for up to 3 months when stored at −20 °C.
  • Protein synonyms:
    Fibroblast Growth Factor-21|FGFL
  • Protein/peptide name:
    FGF21
  • Purity:
    >90% (SDS-PAGE)
  • Sequence:
    1-210
  • Formulation:
    0.2um-filtered solution in PBS.

Specifications

About this item

FGF-21 (Fibroblast growth factor 21) stimulates glucose uptake in differentiated adipocytes via the induction of glucose transporter SLC2A1/GLUT1 expression (but not SLC2A4/GLUT4 expression). Activity requires the presence of betaKlotho.

Fusion-Tag: Fc Tag

This recombinant proteins is for research use only.