Specifications
- Pk:50 µG
- Protein/peptide type:Recombinant
- Species:Mouse
- Tag sequence:C-6 His tag
- Storage conditions:Lyophilized protein should be stored at −20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4-7 °C for 2-7 days. Aliquots of reconstituted samples are stable at −20 °C for 3 months.
- Protein synonyms:CD33|CD33/Myeloid cell surface antigen CD33/Sialic acid-binding Ig-like lectin 3/Siglec-3/Siglec3
- Protein/peptide name:Siglec-3
- Purity:> 95% as determined by reducing SDS-PAGE
- Molecular weight:25.7 kD
- Sequence:Asp18-Glu240
- Formulation:Lyophilized from a 0.2 um filtered solution of 20mM PB,150mM NaCl,pH7.4. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the Lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
- Tested applications:Bioassay
Specifications
About this item
Mouse myeloid cell surface antigen CD33(CD33) is a member of the immunoglobulin superfamily and SIGLEC (sialic acid binding Ig-like lectin) family. CD33 contains one Ig-like C2-type domain and one Ig-like V-type domain. CD33 is a putative adhesion molecule of myelomonocytic-derived cells that mediates sialic-acid dependent binding to cells. CD33 preferentially binds to alpha-2,6-linked sialic acid. The sialic acid recognition site may be masked by cis interactions with sialic acids on the same cell surface. In the immune response, CD33 may act as an inhibitory receptor upon ligand induced tyrosine phosphorylation by recruiting cytoplasmic phosphatase(s) via their SH2 domain(s) that block signal transduction through dephosphorylation of signalling molecules. CD33 induces apoptosis in acute myeloid leukemia. CD33 is becoming increasingly important as a target of antibody-mediated therapy in acute myeloid leukaemia (AML).
Fusion-Tag: C-6 His tag
This recombinant protein can be used for biological assays. For research use only.