Human recombinant FGF1 Intracellular-Binding (from E. coli)
Catalog # PRSI92-063
Supplier: ProSci Inc.
CAS Number:
Specifications
- Fp:50 µG
- Protein/peptide type:Recombinant
- Kilde:E. coli
- Arter:Human
- Lagringsforhold:Store at −20 °C, stable for 6 months after receipt. Please minimize freeze-thaw cycles.
- Protein synonyms:Acidic Fibroblast Growth Factor Intracellular-Binding Protein|FIBP|aFGF Intracellular-Binding Protein|FGF-1 Intracellular-Binding Protein
- Protein/peptide name:FGF1 Intracellular-Binding
- Renhet:> 95% as determined by reducing SDS-PAGE
- Molekylvekt:41.8 kD
- Sekvens:Met1-Asp357
- Formulering:Supplied as a 0.2 um filtered solution of 50mM TrisHCl,pH8.0. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
- Uttestet applikasjon:Bioassay
Specifications
About this item
Acidic Fibroblast Growth Factor Intracellular-Binding Protein (FIBP) is highly expressed in the heart, skeletal muscle, and pancreas. Acidic Fibroblast Growth Factor is a member of the fibroblast growth factor (FGF) family. FGF family members possess broad mitogenic and cell survival activities, and are involved in a variety of biological processes, including embryonic development, cell growth, morphogenesis, tissue repair, tumor growth and invasion. FIBP is an intracellular protein that binds selectively to acidic fibroblast growth factor (aFGF). It is postulated that FIBP may be involved in the mitogenic action of aFGF.
- This recombinant protein can be used for biological assays. For research use only.