Human recombinant LAMP (from cells)
Catalog # PRSI91-862
Supplier: ProSci Inc.
CAS Number:
Specifications
- Pk:50 µG
- Protein/peptide type:Recombinant
- Source:Cells
- Species:Human
- Tag sequence:C-6 His tag
- Storage conditions:Lyophilized protein should be stored at −20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4-7 °C for 2-7 days. Aliquots of reconstituted samples are stable at −20 °C for 3 months.
- Protein synonyms:Limbic system-associated membrane protein|LSAMP|IgLON family member 3
- Protein/peptide name:LAMP
- Purity:> 95% as determined by reducing SDS-PAGE
- Molecular weight:32.8 kD
- Sequence:Val29-Asn315
- Formulation:Lyophilized from a 0.2 um filtered solution of 20mM PB,150mM NaCl,pH7.4. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the Lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
- Tested applications:used for biological assays
Specifications
About this item
Limbic system-associated membrane protein is also known as LSAMP, IgLON family member 3. In humans, it is encoded by the LSAMP gene. It belongs to the immunoglobulin superfamily and contains 3 Ig-like C2-type domains. Limbic system-associated membrane protein mediates selective neuronal growth and axon targeting. It contributes to the guidance of developing axons and remodeling of mature circuits in the limbic system. It is also essential for normal growth of the hyppocampal mossy fiber projection
- This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag