Order Entry
Norway
ContactUsLinkComponent
Human recombinant Cathepsin Z (from Cells)
Human recombinant Cathepsin Z (from Cells)
  PRSI91-394
 :  
Human recombinant Cathepsin Z (from Cells)
  PRSI91-394
 :  

 

  • Pk:
    50 µG
  • Protein/peptide type:
    Recombinant
  • Source:
    Cells
  • Species:
    Human
  • Tag sequence:
    C-6 His tag
  • Storage conditions:
    Store at −20 °C, stable for 6 months after receipt. Please minimize freeze-thaw cycles.
  • Protein synonyms:
    CTSZ|Cathepsin X|Cathepsin Z|Cathepsin P
  • Protein/peptide name:
    Cathepsin Z
  • Purity:
    > 95% as determined by reducing SDS-PAGE
  • Molecular weight:
    32.51 kD
  • Sequence:
    Gly24-Val303
  • Formulation:
    Supplied as a 0.2 um filtered solution of 20mM HAc-NaAc, 150mM NaCl, pH 4.0. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
  • Tested applications:
    Bioassay

 

 

Cathepsin Z is a lysosomal cysteine proteinase and belongs to the peptidase C1 family. Human Cathepsin Z contains a singnal sequence, a propeptide and a mature chain. It exhibits bothcarboxy-monopeptidase and carboxy-dipeptidase activities. In contrast to cathepsin B, it does not act as an endopeptidase. Cathepsin Z is restricted to the cells of theimmune system, predominantly monocytes, macrophages and dendritic cells. It is expressed ubiquitously in cancer cell lines and primary tumors. Like other members of this family, Cathepsin Z may be involved in tumorigenesis.

  • This recombinant protein can be used for biological assays. For research use only.

Fusion-Tag: C-6 His tag