Human recombinant Kallikrein 11 (from Cells)
Catalog # PRSI91-332
Supplier: ProSci Inc.
CAS Number:
Specifications
- Pk:50 µG
- Protein/peptide type:Recombinant
- Source:Cells
- Species:Human
- Tag sequence:C-6 His tag
- Storage conditions:Store at −20 °C, stable for 6 months after receipt. Please minimize freeze-thaw cycles.
- Protein synonyms:Trypsin-like Protease|KLK11|PRSS20|Hippostasin|Serine Protease 20|TLSP|hK11|Kallikrein-11
- Protein/peptide name:Kallikrein 11
- Purity:> 95% as determined by reducing SDS-PAGE
- Molecular weight:26.28 kD
- Sequence:Glu19-Asn250
- Formulation:Supplied as a 0.2 um filtered solution of 20mM TrisHCl, 150mM NaCl, 2mM CaCl2, pH 8.0. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
- Tested applications:Bioassay
Specifications
About this item
Human Kallikrein 11 (KLK11) is a member of tissue kallikrein family which are extracellular serine proteases consisting of 15 members. Two isoforms of KLK11 are differentially expressed. Isoform 1 is predominantly expressed in brain and isoform 2 is preferentially expressed in prostate. Isoform 1 consists of a signal peptide,a short pro peptide and the mature chain.Isoform 2 contains an extra 32 amino acid N terminal to full-length isoform 1.KLK11 is a novel marker for ovarian and prostate cancer carcinomas. KLK11 can be activated by thermolysin and is active against a thioester substrate.
- This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag