Order Entry
Norway
ContactUsLinkComponent
Human recombinant MOB1A (from E. coli)
Human recombinant MOB1A (from E. coli)
Catalog # PRSI91-277
Supplier:  ProSci Inc.
CAS Number:  
Human recombinant MOB1A (from E. coli)
Catalog # PRSI91-277
Supplier:  ProSci Inc.
CAS Number:  

Specifications

  • Pk:
    50 µG
  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Tag sequence:
    C-6 His tag
  • Storage conditions:
    Lyophilized protein should be stored at −20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4-7 °C for 2-7 days. Aliquots of reconstituted samples are stable at −20 °C for 3 months.
  • Protein synonyms:
    MOB4B|Mob1 Homolog 1B|C2orf6|MOB Kinase Activator 1A|Mob1 Alpha|MOBKL1B|Mps One Binder Kinase Activator-like 1B|Mob1A|MOBK1B
  • Protein/peptide name:
    MOB1A
  • Purity:
    > 95% as determined by reducing SDS-PAGE
  • Molecular weight:
    26 kD
  • Sequence:
    Ser2-Arg216
  • Formulation:
    Lyophilized from a 0.2 um filtered solution of 20mM Tris, 0.15M NaCl, pH 8.0 . Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the Lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
  • Tested applications:
    Bioassay

Specifications

About this item

MOB Kinase Activator 1A (MOB1A) belongs to the MOB1/phocein family, which acts as an activator of LATS1/2 in the Hippo signaling pathway, plays a key role in organ size control and tumor suppression by restricting proliferation and promoting apoptosis. MOBKL1B stimulates the kinase activity of STK38 and STK38L. MOBKL1B binds to and regulate downstream targets such as the NDR-family protein kinases and LATS1 kinase.

  • This recombinant protein can be used for biological assays. For research use only.

Fusion-Tag: C-6 His tag