Order Entry
Norway
ContactUsLinkComponent
Human recombinant MOB4 (from E. coli)
Human recombinant MOB4 (from E. coli)
Catalog # PRSI91-266
Supplier:  ProSci Inc.
CAS Number:  
Human recombinant MOB4 (from E. coli)
Catalog # PRSI91-266
Supplier:  ProSci Inc.
CAS Number:  

Specifications

  • Pk:
    50 µG
  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Tag sequence:
    N-6 His tag
  • Storage conditions:
    Lyophilized protein should be stored at −20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4-7 °C for 2-7 days. Aliquots of reconstituted samples are stable at −20 °C for 3 months.
  • Protein synonyms:
    2C4D|Class II mMOB1|MOB4|Preimplantation Protein 3|MOBKL3|PHOCN|Mps One Binder Kinase Activator-like 3|MOB-like Protein Phocein|PREI3|Mob3|Mob1 Homolog 3
  • Protein/peptide name:
    MOB4
  • Purity:
    > 95% as determined by reducing SDS-PAGE
  • Molecular weight:
    28.2 kD
  • Sequence:
    Met1-Ala225
  • Formulation:
    Lyophilized from a 0.2 um filtered solution of 20mM TrisHCl, 150mM NaCl, 1mM DTT, pH 8.0. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the Lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
  • Tested applications:
    For Biological Assays

Specifications

About this item

MOB-Like Protein Phocein is a member of the MOB1/Phocein Family. MOB-Like Protein Phocein is associated with membranes and the Golgi stacks. It is present in the cytosol, where it behaves as a protein complex. It has been shown that MOB-Like Protein Phocein interacts with DNM1, EPS15 and Nucleoside Diphosphate Kinase. MOB-Like Protein Phocein is the major partner of Striatin Family members and may play a important role in membrane trafficking, specifically in membrane budding reactions.

  • This recombinant protein can be used for biological assays. For research use only.

Fusion-Tag: N-6 His tag