Order Entry
Norway
ContactUsLinkComponent
 
Human recombinant LMW-PTP (from E. coli)
  PRSI91-181
 :  
Human recombinant LMW-PTP (from E. coli)
  PRSI91-181
 :  

 

  • Pk:
    50 µG
  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Tag sequence:
    C-6 His tag
  • Storage conditions:
    Store at −20 °C, stable for 6 months after receipt. Please minimize freeze-thaw cycles.
  • Protein synonyms:
    Adipocyte Acid Phosphatase|LMW-PTP|LMW-PTPase|Red Cell Acid Phosphatase 1|Low Molecular Weight Phosphotyrosine Protein Phosphatase|Low Molecular Weight Cytosolic Acid Phosphatase|ACP1
  • Protein/peptide name:
    LMW-PTP
  • Purity:
    > 95% as determined by reducing SDS-PAGE
  • Molecular weight:
    19.04 kD
  • Sequence:
    Ala2-His158
  • Formulation:
    Supplied as a 0.2 um filtered solution of 20mM TrisHCl, 150mM NaCl, 10% Glycerol, pH 8.0. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
  • Tested applications:
    For Biological assays

 

 

Low Molecular Weight Phosphotyrosine Protein Phosphatase (LMW-PTP) is a member of the low molecular weight phosphotyrosine protein phosphatase family. LMW-PTP serves as an acid phosphatase and a protein tyrosine phosphatase (PTPase) by hydrolyzing protein tyrosine phosphate to protein tyrosine and orthophosphate. LMW-PTP can be detected in all human tissues, including adipocytes. LMW-PTP is a cytosolic enzyme that regulate cell proliferation and growth of leiomyomas during dephosphorylation of the PDGF receptor. In addition, LMW-PTP plays an important role in the regulation of physiological functions, such as stress resistance and synthesis of the polysaccharide capsule.

  • This recombinant protein can be used for biological assays. For research use only.

Fusion-Tag: C-6 His tag