Human recombinant BCL-2 Related A1 (from E. coli)
PRSI91-086
:
- Pk:50 µG
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Human
- Tag sequence:C-6 His tag
- Storage conditions:Store at −20 °C, stable for 6 months after receipt. Please minimize freeze-thaw cycles.
- Protein synonyms:Bcl2-L-5|GRS|Bcl-2-like Protein 5|BCL2L5|Protein BFL-1|Bcl-2-Related Protein A1|BFL1|BCL2A1|Protein GRS|HBPA1|Hemopoietic-Specific Early Response Protein
- Protein/peptide name:BCL-2 Related A1
- Purity:> 95% as determined by reducing SDS-PAGE
- Molecular weight:18.52 kD
- Sequence:Met1-Ser152
- Formulation:Supplied as a 0.2 um filtered solution of 20mM PB, 150mM NaCl, 1mM EDTA, 1mM DTT, 10% Glycerol, pH 7.4. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
- Tested applications:Bioassay
Bcl-2-Related Protein A1 (BCL2A1) is a member of of the BCL-2 protein family which act as anti- and pro-apoptotic regulators that are involved in a wide variety of cellular activities such as embryonic development, homeostasis and tumorigenesis. BCL2A1 is a cytoplasm protein and interacts directly with BAK1, BID, BMF and BBC3. BCL2A1 is induced by phorbol ester and inflammatory cytokines, such as TNF or IL1B/interleukin-1 beta, but not by growth factors. BCL2A1 is invovled in the response of hemopoietic cells to external signals and in maintaining endothelial survival during infection.
- This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag