Order Entry
Norway
ContactUsLinkComponent
 
Human recombinant IL7 (from E. coli)
  PRSI91-070
 :  
Human recombinant IL7 (from E. coli)
  PRSI91-070
 :  

 

  • Pk:
    50 µG
  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Storage conditions:
    Lyophilized protein should be stored at −20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4-7 °C for 2-7 days. Aliquots of reconstituted samples are stable at −20 °C for 3 months.
  • Protein synonyms:
    IL-7|IL7|Interleukin-7
  • Protein/peptide name:
    IL7
  • Purity:
    > 95% as determined by reducing SDS-PAGE
  • Molecular weight:
    17.5 kD
  • Sequence:
    Asp26-His177
  • Formulation:
    Lyophilized from a 0.2 um filtered solution of 20mM PB, 250mM NaCl, pH 7.2. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the Lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
  • Tested applications:
    Bioassay

 

 

Human Interleukin 7 (IL-7) is a potent lymphoid cell growth factor stimulating the proliferation of lymphoid progenitors. IL7 can associate with the hepatocyte growth factor (HGF) to form a hybrid cytokine that functions as a pre-pro-B cell growth-stimulating factor. Human IL7 cDNA encodes a 177 amino acid precursor protein containing a 25 amino acid signal peptide and a 152 amino acid mature protein. Human and mouse IL7 share 65% sequence identity in the mature region and both exhibit cross-species activity. IL-7 signals via IL-7 receptor (IL7R) activating multiple pathways including JaK/STAT and PI3K/AKT, which regulate lymphocyte survival, glucose uptake, proliferation, and differentiation. IL-7 is also associated with cytoplasmic IL2-R gamma for signal transduction.

  • This recombinant protein can be used for biological assays. For research use only.