Human recombinant Ror 2 (from HEK293 cells)
PRSI96-652
:
- Pk:50 µG
- Protein/peptide type:Recombinant
- Source:HEK293 cells
- Species:Human
- Protein synonyms:NTRKR2|ROR2
- Protein/peptide name:Ror 2
- Purity:>95% as determined by SDS-PAGE.
- Molecular weight:68 kDa
- Formulation:Lyophilized, 50 mM Tris, 100 mM NaCl, pH 7.5
- Tested applications:Western Blot
Tyrosine-protein kinase transmembrane receptor ROR2 is also known as Neurotrophic tyrosine kinase, receptor-related 2 (NTRKR2), which belongs to the protein kinase superfamily and Tyr protein kinase family and ROR subfamily. ROR2 is a homodimer protein, which can binds YWHAB or interact with WTIP. ROR2 may be involved in the early formation of the chondrocytes. It seems to be required for cartilage and growth plate development.