Order Entry
Norway
ContactUsLinkComponent
Human recombinant Ror 1 (from HEK293 cells)
Human recombinant Ror 1 (from HEK293 cells)
Catalog # PRSI96-651
Supplier:  ProSci Inc.
CAS Number:  
undefined
Human recombinant Ror 1 (from HEK293 cells)
Catalog # PRSI96-651
Supplier:  ProSci Inc.
CAS Number:  

Specifications

  • Pk:
    100 µG
  • Protein/peptide type:
    Recombinant
  • Source:
    HEK293 cells
  • Species:
    Human
  • Protein synonyms:
    NTRKR1|ROR1
  • Protein/peptide name:
    Ror 1
  • Purity:
    >95% as determined by SDS-PAGE.
  • Molecular weight:
    68.4 kDa
  • Formulation:
    Lyophilized, 50 mM tris, 100 mM glycine, pH7.5
  • Tested applications:
    Western Blot

Specifications

About this item

Tyrosine-protein kinase transmembrane receptor ROR1 is also known as Neurotrophic tyrosine kinase, receptor-related 1 (NTRKR1), which belongs to the protein kinase superfamily or tyr protein kinase family or ROR subfamily. ROR1 contains 1 FZ (frizzled) domain, 1 Ig-like C2-type (immunoglobulin-like) domain, 1 kringle domain, 1 protein kinase domain. ROR1 is expressed at high levels during early embryonic development. The expression levels drop strongly around day 16 and there are only very low levels in adult tissues. Isoform Short is strongly expressed in fetal and adult CNS and in a variety of human cancers, including those originating from CNS or PNS neuroectoderm. ROR1 could interact with casein kinase 1 epsilon (CK1ε) to activate phosphoinositide 3-kinase-mediated AKT phosphorylation and cAMP-response-element-binding protein (CREB), which was associated with enhanced tumor-cell growth.