Order Entry
Norway
ContactUsLinkComponent
Human recombinant MMP9 (catalytic domain) (from HEK293 cells)
Human recombinant MMP9 (catalytic domain) (from HEK293 cells)
Catalog # PRSI96-539
Supplier:  ProSci Inc.
CAS Number:  
undefined
Human recombinant MMP9 (catalytic domain) (from HEK293 cells)
Catalog # PRSI96-539
Supplier:  ProSci Inc.
CAS Number:  

Specifications

  • Pk:
    50 µG
  • Protein/peptide type:
    Recombinant
  • Source:
    HEK293 cells
  • Species:
    Human
  • Biological activity:
    Measured by its ability to cleave the fluorogenic peptide substrate, Mca-PLGL-Dpa-AR-NH2. The specific activity is >1200 pmol/min/μg.
  • Protein synonyms:
    GELB|MMP9|MANDP2|Gelatinase B|CLG4B
  • Protein/peptide name:
    MMP9 (catalytic domain)
  • Purity:
    >95% as determined by SDS-PAGE.
  • Molecular weight:
    50.8 kDa
  • Formulation:
    Lyophilized, PBS, pH7.4
  • Tested applications:
    Western Blot

Specifications

About this item

Matrix metallopeptidase 9 (MMP-9) is also known as 92 kDa type IV collagenase, 92 kDa gelatinase or gelatinase B (GELB), CLG4B, is secreted from neutrophils, macrophages, and a number of transformed cells, and is the most complex family member in terms of domain structure and regulation of its activity. . Structurally, MMP9 maybe be divided into five distinct domains: a prodomain which is cleaved upon activation, a gelatinbinding domain consisting of three contiguous fibronectin type II units, a catalytic domain containing the zinc binding site, a prolinerich linker region, and a carboxyl terminal hemopexinlike domain. This enzyme degrades various substrates including gelatin, collagen types IV and V, and elastin. MMP9 is involved in a variety of autoimmune diseases such as systemic lupus erythematosus, rheumatoid arthritis, and multiple sclerosis, and be regarded as a potential therapeutic target.