Human recombinant IFNAR2 (from HEK293 cells)
Catalog # PRSI96-408
Supplier: ProSci Inc.
CAS Number:
Specifications
- Pk:100 µG
- Protein/peptide type:Recombinant
- Source:HEK293 cells
- Species:Human
- Protein synonyms:IFNARB|IFN-alpha/beta receptor 2|IFNABR|IFNAR2|IFN-R-2
- Protein/peptide name:IFNAR2
- Purity:>95% as determined by SDS-PAGE.
- Molecular weight:51.4 kDa
- Formulation:Lyophilized, 50 mM tris, 100 mM glycine, pH7.5
- Tested applications:Western Blot