Human SAPAP4 peptide (13 aa in the center)
Catalog # PRSI4633P
Supplier: ProSci Inc.
CAS Number:
Specifications
- Pk:50 µG
- Species:Human
- Storage conditions:Store at –20 °C, stable for one year.
- Amino acid number:13 aa in the center
- Protein/peptide name:SAPAP4 Peptide (13 aa in the center)
- Tested applications:Blocking
Specifications
About this item
SAPAP4 peptide is used for blocking the activity of SAPAP4 antibody.