Order Entry
Norway
ContactUsLinkComponent
 
Human CTRP6 peptide (15 aa in the center)
  PRSI3577P
 :  
Human CTRP6 peptide (15 aa in the center)
  PRSI3577P
 :  

 

  • Pk:
    50 µG
  • Species:
    Human
  • Storage conditions:
    Store at –20 °C, stable for one year.
  • Amino acid number:
    15 aa in the center
  • Protein/peptide name:
    CTRP6 Peptide (15 aa in the center)
  • Tested applications:
    Blocking

 

 

CTRP6 peptide is used for blocking the activity of CTRP6 antibody.