Anti-CCL24 Mouse Monoclonal Antibody [clone: 1.1_6C7-2C5]
PRSIXP-5715-M
- Antibody type:Primary
- Antigen name:Chemokine (C-C motif) ligand 24
- Antigen symbol:CCL24
- Clonality:Monoclonal
- Clone:1.1_6C7-2C5
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Reactivity:Human
- Western blot:Yes
- Form:Lyophilized
- Gene ID:6369
- Antigen synonyms:C-C motif chemokine 24|Ckb-6|MPIF-2|MPIF2|SCYA24|CK-beta-6
- Storage temperature:Stable at room temperature for at least one month and for greater than a year when kept at –20 °C. Reconstitute in sterile water to a concentration of 0.1 - 1.0 mg/mL. Stable for at least six weeks at 2-4 °C when reconstituted.
- Shipping temperature:RT
- Immunogen:The monoclonal antibody was produced in BALB/c X ICR F1 mice using recombinant human Eotaxin-2 as the immunizing antigen.
- Purification:This IgG1K antibody was purified from ascites fluid followed by Protein A affinity chromatography.
- Size:500 µg
- Pk:500 µG
Type: Primary
Antigen: CCL24
Clonality: Monoclonal
Clone: 1.1_6C7-2C5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human