Anti-IL8 Mouse Monoclonal Antibody [clone: SB-84-8]
Catalog # PRSIXP-5706-M
Supplier: ProSci Inc.
Specifications
- Antibody type:Primary
- Antigen name:Interleukin 8
- Antigen symbol:IL8
- Clonality:Monoclonal
- Clone:SB-84-8
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Reactivity:Human
- Western blot:Yes
- Form:Lyophilized
- Gene ID:3576
- Antigen synonyms:MDNCF|Interleukin-8|T-cell chemotactic factor|LYNAP|NAP1|NAP-1|GCP1|Monocyte-derived neutrophil chemotactic factor|MONAP|LECT|NAF|GCP-1|CXCL8|LUCT|Neutrophil-activating protein 1
- Storage temperature:Stable at room temperature for at least one month and for greater than a year when kept at –20 °C. Reconstitute in sterile water to a concentration of 0.1 - 1.0 mg/mL. Stable for at least six weeks at 2-4 °C when reconstituted.
- Shipping temperature:RT
- Immunogen:The monoclonal antibody was produced in BALB/c x C57BL/6 F1 mice using recombinant hIL-8 as the immunizing antigen.
- Purification:This IgG1 antibody was purified from ascites fluid on a Protein A affinity column.
- Size:500 µg
- Pk:500 µG
Specifications
About this item
IL-8 is a member of the CXC chemokine family. This family of small basic heparan-binding proteins are proinflammatory and primarily mediate the activation and migration of neutrophils into tissue from peripheral blood. This chemokine is one of the major mediators of the inflammatory response and is secreted by several cell types in response to an inflammatory stimulus. It functions as a chemoattractant, and is also a potent angiogenic factor. IL-8 attracts neutrophils, basophils, and T-cells, but not monocytes.
Type: Primary
Antigen: IL8
Clonality: Monoclonal
Clone: SB-84-8
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human