Anti-IL9 Rabbit Polyclonal Antibody (Biotin)
Catalog # PRSIXP-5200BT
Supplier: ProSci Inc.
Specifications
- Antigen name:Interleukin 9
- Antigen symbol:IL9
- Clonality:Polyclonal
- Conjugation:Biotin
- ELISA:Yes
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Form:Lyophilized
- Antigen synonyms:Interleukin-9|IL-9|IL-9 |Cytokine P40|HP40|P40
- Immunogen:Produced from sera of rabbits pre-immunized with highly pure (>98%) recombinant hIL-9 (human Interleukin-9).
- Size:50 µg
- Pk:50 µG
Specifications
About this item
IL8 is a member of the CXC chemokine family. This family of small basic heparan-binding proteins are proinflammatory and primarily mediate the activation and migration of neutrophils into tissue from peripheral blood.
Anti-hIL-9 specific antibody was purified by affinity chromatography and then biotinylated.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type:
Antigen: IL9
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human