Anti-MADCAM1 Mouse Monoclonal Antibody
Katalog # PRSI49-044
Leverandør: ProSci Inc.
Spesifikasjoner
- Antigen name:Mucosal Vascular Addressin Cell Adhesion Molecule 1
- Antigen symbol:MADCAM1
- Clonality:Monoclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1
- Reactivity:Human
- Western blot:Yes
- Form:Liquid
- Antigen synonyms:mucosal addressin cell adhesion molecule-1|mucosal addressin cell adhesion molecule 1|MACAM1|hMAdCAM-1|MADCAM-1
- Immunogen:Recombinant soluble MAdCAM-1-fc fusion protein
- Tested applications:Fluorescence-Activated Cell Sorting
- Size:50 µg
- Pk:100 µG
Spesifikasjoner
Om dette produktet
Protein G Column
PBS, pH 7.4.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type:
Antigen: MADCAM1
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human