Order Entry
Norway
ContactUsLinkComponent
Anti-CDT1 Goat Polyclonal Antibody
Anti-CDT1 Goat Polyclonal Antibody
Catalog # PRSI45-386
Supplier:  ProSci Inc.
undefined
Anti-CDT1 Goat Polyclonal Antibody
Catalog # PRSI45-386
Supplier:  ProSci Inc.

Specifications

  • Antigen name:
    Chromatin Licensing And Dna Replication Factor 1
  • Antigen symbol:
    CDT1
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Goat
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Form:
    Liquid
  • Antigen synonyms:
    DUP|RIS2
  • Concentration:
    500 ug/mL
  • Immunogen:
    CDT1 antibody was raised against a 13 amino acid synthetic peptide near the C-Terminus of CDT1.
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

CDT1 antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.

CDT1 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: CDT1
Clonality: Polyclonal
Clone: 0
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: