Anti-FUT1 Goat Polyclonal Antibody
Catalog # PRSI42-384
Supplier: ProSci Inc.
Specifications
- Antigen name:Fucosyltransferase 1
- Antigen symbol:FUT1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- Reactivity:Rat,Mouse
- Western blot:Yes
- Form:Liquid
- Antigen synonyms:GDP-L-fucose:beta-D-galactoside 2-alpha-L-fucosyltransferase 1|Alpha(1,2)FT 1|Blood group H alpha 2-fucosyltransferase|Galactoside 2-alpha-L-fucosyltransferase 1|FUT1|Fucosyltransferase 1|H|HSC
- Concentration:500 ug/mL
- Immunogen:Fut1 antibody was raised against a amino acid synthetic peptide near the internal region of Fut1 (mouse).
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Fut1 antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.
Fut1 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.
Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.
Type:
Antigen: Fut1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Mouse, Rat