Anti-PSP Goat Polyclonal Antibody
PRSI42-343
- Antigen name:Prostate Secretory Protein
- Antigen symbol:PSP
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- Form:Liquid
- Antigen synonyms:Microseminoprotein|beta-
- Concentration:500 ug/mL
- Immunogen:Psp antibody was raised against a 15 amino acid synthetic peptide near the internal region of Psp.
- Size:100 µg
- Pk:100 µG
Psp antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.
Psp antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.
Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.
Type:
Antigen: PSP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: 0