Anti-NGF Mouse Monoclonal Antibody
PRSI38-239
- Antigen name:Nerve Growth Factor
- Antigen symbol:NGF
- Clonality:Monoclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Reactivity:Human
- Western blot:Yes
- Neutralisation:Yes
- Form:Lyophilized
- Antigen synonyms:HSAN5|Beta-NGF|NGFB
- Immunogen:beta-NGF antibody is produced in mice using highly pure (>98%) recombinant human beta-NGF as the immunizing antigen.
- Size:500 µg
- Pk:500 µG
This IgG2bK antibody was purified from cell culture by Protein G affinity chromatography.
The reconstituted antibody can temporarily be stored at 2-8°C. Long term storage at -20°C or below. Avoid repeated freeze-thaw cycles.
Type:
Antigen: NGF
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human