Order Entry
Norway
ContactUsLinkComponent
Anti-PMCH Rabbit Polyclonal Antibody
Anti-PMCH Rabbit Polyclonal Antibody
Catalog # PRSI31-122
Supplier:  ProSci Inc.
undefined
Anti-PMCH Rabbit Polyclonal Antibody
Catalog # PRSI31-122
Supplier:  ProSci Inc.

Specifications

  • Antigen name:
    Pro-Melanin Concentrating Hormone
  • Antigen symbol:
    PMCH
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Form:
    Lyophilized
  • Antigen synonyms:
    MCH
  • Immunogen:
    A synthetic peptide corresponding a region of human PMCH.
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

MCH may act as a neurotransmitter or neuromodulator in a broad array of neuronal functions directed toward the regulation of goal-directed behavior, such as food intake, and general arousal. May also have a role in spermatocyte differentiation.

Antibody is purified by Protein A/G affinity chromatography method.

Antibody is lyophilized in PBS buffer with 2% sucrose. Add 100 μL of distilled water. Final antibody concentration is 1 mg/mL.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: PMCH
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human