Order Entry
Norway
ContactUsLinkComponent
Anti-HTR7 Rabbit Polyclonal Antibody
Anti-HTR7 Rabbit Polyclonal Antibody
Catalog # PRSI31-119
Supplier:  ProSci Inc.
undefined
Anti-HTR7 Rabbit Polyclonal Antibody
Catalog # PRSI31-119
Supplier:  ProSci Inc.

Specifications

  • Antigen name:
    5-Hydroxytryptamine (serotonin) Receptor 7 (adenylate cyclase-coupled)
  • Antigen symbol:
    HTR7
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Form:
    Lyophilized
  • Antigen synonyms:
    Serotonin receptor 7|5-HT7,5-HT-X
  • Immunogen:
    A synthetic peptide corresponding a region of human HTR7.
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

This is one of the several different receptors for 5- hydroxytryptamine (serotonin), a biogenic hormone that functions as a neurotransmitter, a hormone, and a mitogen. The activity of this receptor is mediated by g proteins that stimulate adenylate cyclase.

Antibody is lyophilized in PBS buffer with 2% sucrose. Add 100 μL of distilled water. Final antibody concentration is 1 mg/mL.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: HTR7
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human