Anti-UNC93B1 Rabbit Polyclonal Antibody
Catalog # PRSI30-564
Supplier: ProSci Inc.
Specifications
- Antigen name:Unc-93 homolog B1
- Antigen symbol:UNC93B1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Form:Lyophilized
- Antigen synonyms:Unc-93B1|Protein unc-93 homolog B1|UNC93B|IIAE1|UNC93|Unc93b
- Immunogen:A synthetic peptide corresponding a region of human UNC93B1.
- Size:100 µg
- Pk:100 µG
Specifications
About this item
UNC93B1 is a protein with similarity to the C. elegans unc93 protein. The Unc93 protein is involved in the regulation or coordination of muscle contraction in the worm.This gene encodes a protein with similarity to the C. elegans unc93 protein. The Unc93 protein is involved in the regulation or coordination of muscle contraction in the worm.
Antibody is purified by protein A chromatography method.
Antibody is lyophilized in PBS buffer with 2% sucrose. Add 100 μL of distilled water. Final antibody concentration is 1 mg/mL.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type:
Antigen: UNC93B1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat