Anti-KLF17 Rabbit Polyclonal Antibody
Catalog # PRSI25-574
Supplier: ProSci Inc.
Specifications
- Antigen name:Kruppel-like Factor 17
- Antigen symbol:KLF17
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Form:Lyophilized
- Antigen synonyms:zinc finger protein 393|ZNF393|Kruppel-like factor 17|Zfp393
- Immunogen:A synthetic peptide corresponding a region of human KLF17.
- Size:50 µg
- Pk:50 µG
Specifications
About this item
KLF17 binds G/C-rich sites via its zinc fingers and activates transcription from CACCC-box elements. It may be a germ cell-specific transcription factor that plays important roles in spermatid differentiation and oocyte development.
Antibody is purified by peptide affinity chromatography method.
Antibody is lyophilized in PBS buffer with 2% sucrose. Add 50 μL of distilled water. Final antibody concentration is 1 mg/mL.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type:
Antigen: KLF17
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human