Order Entry
Norway
ContactUsLinkComponent
Anti-TNFSF12 Rabbit Polyclonal Antibody
Anti-TNFSF12 Rabbit Polyclonal Antibody
Catalog # PRSI25-364
Supplier:  ProSci Inc.
undefined
Anti-TNFSF12 Rabbit Polyclonal Antibody
Catalog # PRSI25-364
Supplier:  ProSci Inc.

Specifications

  • Antigen name:
    Tumor Necrosis Factor Superfamily Member 12
  • Antigen symbol:
    TNFSF12
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Form:
    Lyophilized
  • Antigen synonyms:
    UNQ181/PRO207|APO3L|APO3 ligand|TWEAK|Tumor necrosis factor ligand superfamily member 12|DR3LG
  • Immunogen:
    A synthetic peptide corresponding a region of human TNFSF12.
  • Size:
    50 µg
  • Pk:
    50 µG

Specifications

About this item

TNFSF12 is a cytokine that belongs to the tumor necrosis factor (TNF) ligand family. This protein is a ligand for the FN14/TWEAKR receptor. This cytokine has overlapping signaling functions with TNF, but displays a much wider tissue distribution. This cytokine can induce apoptosis via multiple pathways of cell death in a cell type-specific manner. This cytokine is also found to promote proliferation and migration of endothelial cells, and thus acts as a regulator of angiogenesis. Two alternatively spliced transcript variants of this gene encoding distinct isoforms have been reported. The protein encoded by this gene is a cytokine that belongs to the tumor necrosis factor (TNF) ligand family. This protein is a ligand for the FN14/TWEAKR receptor. This cytokine has overlapping signaling functions with TNF, but displays a much wider tissue distribution. This cytokine can induce apoptosis via multiple pathways of cell death in a cell type-specific manner. This cytokine is also found to promote proliferation and migration of endothelial cells, and thus acts as a regulator of angiogenesis. Publication Note: This RefSeq record includes a subset of the publications that are available for this gene. Please see the Entrez Gene record to access additional publications.

Antibody is purified by peptide affinity chromatography method.

Antibody is lyophilized in PBS buffer with 2% sucrose. Add 50 μL of distilled water. Final antibody concentration is 1 mg/mL.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: TNFSF12
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human