Order Entry
Norway
ContactUsLinkComponent
Anti-DUSP12 Rabbit Polyclonal Antibody
Anti-DUSP12 Rabbit Polyclonal Antibody
Catalog # PRSI25-098
Supplier:  ProSci Inc.
undefined
Anti-DUSP12 Rabbit Polyclonal Antibody
Catalog # PRSI25-098
Supplier:  ProSci Inc.

Specifications

  • Antigen name:
    Dual Specificity Phosphatase 12
  • Antigen symbol:
    DUSP12
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Form:
    Lyophilized
  • Antigen synonyms:
    YVH1|serine/threonine specific protein phosphatase|dual specificity protein phosphatase 12|DUSP1|dual specificity tyrosine phosphatase YVH1|YVH1 protein-tyrosine phosphatase ortholog
  • Immunogen:
    A synthetic peptide corresponding a region of human DUSP12.
  • Size:
    50 µg
  • Pk:
    50 µG

Specifications

About this item

DUSP12 is a member of the dual specificity protein phosphatase subfamily. These phosphatases inactivate their target kinases by dephosphorylating both the phosphoserine/threonine and phosphotyrosine residues. They negatively regulate members of the mitogen-activated protein (MAP) kinase superfamily (MAPK/ERK, SAPK/JNK, p38), which is associated with cellular proliferation and differentiation. Different members of the family of dual specificity phosphatases show distinct substrate specificities for various MAP kinases, different tissue distribution and subcellular localization, and different modes of inducibility of their expression by extracellular stimuli. DUSP12 is the human ortholog of the Saccharomyces cerevisiae YVH1 protein tyrosine phosphatase. It is localized predominantly in the nucleus, and is novel in that it contains, and is regulated by a zinc finger domain.The protein encoded by this gene is a member of the dual specificity protein phosphatase subfamily. These phosphatases inactivate their target kinases by dephosphorylating both the phosphoserine/threonine and phosphotyrosine residues. They negatively regulate members of the mitogen-activated protein (MAP) kinase superfamily (MAPK/ERK, SAPK/JNK, p38), which is associated with cellular proliferation and differentiation. Different members of the family of dual specificity phosphatases show distinct substrate specificities for various MAP kinases, different tissue distribution and subcellular localization, and different modes of inducibility of their expression by extracellular stimuli. This gene product is the human ortholog of the Saccharomyces cerevisiae YVH1 protein tyrosine phosphatase. It is localized predominantly in the nucleus, and is novel in that it contains, and is regulated by a zinc finger domain.

DUSP12 antibody can be used for detection of DUSP12 by ELISA at 1:1562500. DUSP12 antibody can be used for detection of DUSP12 by western blot at 1 ?g/mL, and HRP conjugated secondary antibody should be diluted 1:50,000 - 100,000.

Antibody is purified by peptide affinity chromatography method.

Antibody is lyophilized in PBS buffer with 2% sucrose. Add 50 ?L of distilled water. Final antibody concentration is 1 mg/mL.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: DUSP12
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat