Order Entry
Norway
ContactUsLinkComponent
Anti-GITRL Rabbit Polyclonal Antibody
Anti-GITRL Rabbit Polyclonal Antibody
Catalog # PRSI3591
Supplier:  ProSci Inc.
undefined
Anti-GITRL Rabbit Polyclonal Antibody
Catalog # PRSI3591
Supplier:  ProSci Inc.

Specifications

  • Antigen name:
    Tumor Necrosis Factor (Ligand) Superfamily, Member 18
  • Antigen symbol:
    GITRL
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Mouse
  • Western blot:
    Yes
  • Antigen synonyms:
    hGITRL|TL6|AITRL|GITRL
  • Storage buffer:
    PBS containing 0.02% sodium azide
  • Immunogen:
    GITRL antibody was raised against purified recombinant human GITR ligand.
  • Tested applications:
    E, WB, ICC
  • Purification:
    Affinity chromatography purified via peptide column
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

GITRL Antibody: The tumor necrosis factor (TNF) and TNF receptor (TNFR) gene superfamilies regulate numerous biological functions including cell proliferation, differentiation, and survival through regulating the activation of the transcription factor NF-kappa B and various mitogen-activated protein kinases. The glucocorticoid-induced tumor necrosis factor receptor (GITR) is an emerging member of this family that is expressed on CD4+ CD25+ regulatory T cells and appears to have crucial immune regulation functions. Its ligand GITRL is expressed in endothelial and antigen-presenting cells and can activate NF-kappa B, induce both pro- and anti-apoptotic effects, inhibit the suppressive activity of regulatory T cells, and co-stimulate responder T cells through GITR. Dominant negative forms of NIK and TRAF2 expressed in transfected 293 cells substantially inhibited NF-kappa B activation, suggesting that the GITRL-GITR pathway involves both NIK and TRAF2.

Type:
Antigen: GITRL
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse