Anti-Fabp4 Rabbit Polyclonal Antibody
BSBTA01528
- Antibody type:Primary
- Antigen name:Fatty acid-binding protein, adipocyte
- Antigen symbol:Fabp4
- Clonality:Polyclonal
- ELISA:Yes
- Host:Rabbit
- Reactivity:Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Rabbit IgG polyclonal antibody for Fatty acid-binding protein, adipocyte(Fabp4) detection. Tested with WB, ELISA in Mouse;Rat.
Type: Primary
Antigen: Fabp4
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Mouse;Rat