Anti-DDX3X Rabbit Polyclonal Antibody
BSBTA00751-1
- Antibody type:Primary
- Antigen name:ATP-dependent RNA helicase DDX3X
- Antigen symbol:DDX3X
- Clonality:Polyclonal
- Host:Rabbit
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Rabbit IgG polyclonal antibody for ATP-dependent RNA helicase DDX3X(DDX3X) detection. Tested with WB in Human;Mouse;Rat.
Type: Primary
Antigen: DDX3X
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat