Order Entry
Norway
ContactUsLinkComponent
 
Anti-HFE Rabbit Polyclonal Antibody
  BSBTA00506-1
Anti-HFE Rabbit Polyclonal Antibody
  BSBTA00506-1

 

  • Antibody type:
    Primary
  • Antigen name:
    Hereditary hemochromatosis protein
  • Antigen symbol:
    HFE
  • Clonality:
    Polyclonal
  • Host:
    Rabbit
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Size:
    100 µg
  • Pk:
    100 µG

 

 

Rabbit IgG polyclonal antibody for Hereditary hemochromatosis protein(HFE) detection. Tested with WB in Human.

Type: Primary
Antigen: HFE
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human