Anti-ERBB2 Rabbit Polyclonal Antibody
Catalog # BSBTA00010-2
Supplier: Boster Bio
Specifications
- Antibody type:Primary
- Antigen name:Receptor tyrosine-protein kinase erbB-2
- Antigen symbol:ERBB2
- Clonality:Polyclonal
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human,Rat
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Rabbit IgG polyclonal antibody for Receptor tyrosine-protein kinase erbB-2(ERBB2) detection. Tested with WB, IHC-P in Human;Rat.
Type: Primary
Antigen: ERBB2
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Rat