Order Entry
Norway
ContactUsLinkComponent
Anti-CRX Rabbit Polyclonal Antibody
Anti-CRX Rabbit Polyclonal Antibody
Catalog # BSBTA02202-1
Supplier:  Boster Bio
Anti-CRX Rabbit Polyclonal Antibody
Catalog # BSBTA02202-1
Supplier:  Boster Bio

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Cone-rod homeobox protein
  • Antigen symbol:
    CRX
  • Clonality:
    Polyclonal
  • Host:
    Rabbit
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

Rabbit IgG polyclonal antibody for Cone-rod homeobox protein(CRX) detection. Tested with WB in Human.

Type: Primary
Antigen: CRX
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human