Anti-AMH Rabbit Polyclonal Antibody
BSBTA00763
- Antibody type:Primary
- Antigen name:Muellerian-inhibiting factor(
- Antigen symbol:AMH
- Clonality:Polyclonal
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Rabbit IgG polyclonal antibody for Muellerian-inhibiting factor(AMH) detection. Tested with WB in Human.
Type: Primary
Antigen: AMH
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human