Order Entry
Norway
ContactUsLinkComponent
Organic reference standards, trimebutines, Ph. Eur., RS
Organic reference standards, trimebutines, Ph. Eur., RS
Supplier:  EDQM
Certificates
Organic reference standards, trimebutines, Ph. Eur., RS
Supplier:  EDQM

Specifications

  • CAS No.:
    34140-59-5
  • Formula:
    C₂₆H₃₃NO₉
  • Description:
    Trimebutine for system suitability

Specifications