Order Entry
Norway
ContactUsLinkComponent
Organic reference standards, mesalazine, Ph. Eur. RS
Organic reference standards, mesalazine, Ph. Eur. RS
Supplier:  EDQM
Certificates
Organic reference standards, mesalazine, Ph. Eur. RS
Supplier:  EDQM
Certificates

Specifications

  • CAS No.:
    95-55-6
  • Formula:
    C₆H₇NO
  • Description:
    Mesalazine impurity C

Specifications

Related Information