Order Entry
Norway
ContactUsLinkComponent
 
Organic reference standards, lovastatins, Ph. Eur.
 :  EDQM
 
Organic reference standards, lovastatins, Ph. Eur.
 :  EDQM

 

  • CAS No.:
    75330-75-5
  • Formula:
    C₂₄H₃₆O₅
  • Description:
    Lovastatin for peak identification